Acetonitrile, anhydrous ≥99.9% (by GC), OmniSolv® for biochemistry, Supelco®

Supplier: MilliporeSigma
Danger

AX0151-6 AX0151-NP20 AX0151-1 AX0151P-1
CAAX0151-36EA 23932.6 CAD
CAAX0151-36 CAAX0151-37 CAAX0151-38 CAAX0151-40 CAAX0151-6 CAAX0151-NP20 CAAX0151-1 CAAX0151P-1
Acetonitrile, anhydrous ≥99.9% (by GC), OmniSolv® for biochemistry, Supelco®
Acetonitrile

Biosynthesis

Formula: H₃CCN
Boiling Pt: 81…82 °C (lit.)
Melting Pt: –45 °C (lit.)
Density: 0.786 g/ml (25°C) (lit.)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Free amine (as dimethylamine) 0.20ppm max
Gradient at 210 nm 0.003AU max
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
Titrable acid 0.8µeq/g max
Titrable base 0.16µeq/g max
UV Abs. at 189 nm 1.00AU max
UV Abs. at 195 nm 0.15AU max
UV Abs. at 210 nm 0.03AU max
UV Abs. at 230 nm 0.005AU max
UV Abs. at 250 nm 0.005AU max
UV Abs. at 270 nm 0.005AU max
UV Cut-off 189nm max
Water (H2O) 10ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR