Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
AX0116-NP20
AX0116-1
AX0116-6
AX0116P-1
CAAX0116-NP20EA
1834
CAD
CAAX0116-NP20
CAAX0116-1
CAAX0116-6
CAAX0116P-1
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Acetone
For spectrophotometry, chromatography, and residue analysis.
Conforms to IR-spectrum identity. Filtered through 0.2µm filter.
Certifications: Certificate of lot analysis on label.
Caution: Flammable with toxic effects.
Formula:
CH₃COCH₃ Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
Assay (GC, corrected for water content) | 99.5% min |
Capillary ECD responsive substances (as PCNB) | 2ng/L max |
Capillary FID responsive substances (as decane) | 3µg/L max |
Color (APHA) | 10 max |
Residue after evaporation | 1ppm max |
Filtered through 0.2 µm filter | To pass test |
Fluorescence (as quinine base) | 1.0ppb max |
Infrared spectrum | To Pass Test |
Titrable acid | 0.3µeq/g max |
UV Abs. at 329 nm | 1.00AU max |
UV Abs. at 330 nm | 0.80AU max |
UV Abs. at 335 nm | 0.30AU max |
UV Abs. at 340 nm | 0.06AU max |
UV Abs. at 350 nm | 0.005AU max |
UV Abs. at 400 nm | 0.005AU max |
UV Cut-off | 329nm max |
Water (H2O) | 0.5% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...