Ammonium sulfate, OmniPur® for biotechnology, Millipore®

Supplier: MilliporeSigma

2150-1KG 2160-5KG
CA-EM2150EA 570.36 CAD
CA-EM2150 CA-EM2160
Ammonium sulfate, OmniPur® for biotechnology, Millipore®
Ammonium sulphate

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: (NH₄)₂SO₄
Melting Pt: >280 °C (dec., lit.)
Density: 1.77 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR