Ammonium acetate ≥97.0%, OmniPur®, Millipore®
Supplier: MilliporeSigma
|
Warning
|
2145-1KG
2149-5KG
CAEM2145EA
609.46
CAD
CAEM2145
CAEM2149
Ammonium acetate ≥97.0%, OmniPur®, Millipore®
Ammonium acetate
For molecular biology. DNase-, RNase-, and protease-free.
Formula:
H₃CCOONH₄ Melting Pt: 114 °C Density: 1.17 g/cm³ (20 °C) Flash Pt: 136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 EINECS: 211-162-9 Merck Index: 13,00495 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...