Acetonitrile ≥99.9%, B&J Brand™ ACS, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
015-1L
015-4
NP015-20
NS015-20
CABJ015-1LCS
991.58
CAD
CABJ015-1L
CABJ015-4
CABJNP015-20
BJNS015-20
Acetonitrile ≥99.9%, B&J Brand™ ACS, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™
Acetonitrile
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
H₃CCN Boiling Pt: 82 °C (1.013 hPa) Melting Pt: –46 °C Density: 0.7822 g/cm³ (20 °C) Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Electron Capture GC | 10 ng/L max. |
Fluorescence (as quinine base) | 0.3 ppb max. |
GC Analysis | 99.9 % min. |
HPLC Gradient @ 215 nm | 0.005 AU max. |
HPLC Gradient @ 254 nm | 0.001 AU max. |
Refractive Index (20ºC) | 1.3434 - 1.3446 |
Residue | 1 mg/L max. |
UV Cutoff | 190 nm max. |
Water by Karl Fischer Titration | 0.01 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...