Acetone ≥99.9% (dried basis), B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

010-1L 010-4 NP010-20
CABJ010-1LCS 523.44 CAD
CABJ010-1L CABJ010-4 CABJNP010-20 BJNS010-20
Acetone ≥99.9% (dried basis), B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
Acetone

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
Water by Karl Fischer Titration 0.5 % max.
GC Analysis (excluding water) 99.9 % min.
Refractive Index (20ºC) 1.3583 - 1.3589
Residue 1 mg/L max.
UV Absorbance @ 340 nm 0.06 AU max.
UV Absorbance @ 350 nm 0.01 AU max.
UV Absorbance @ 375 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 330 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR