KIMBLE® KONTES® Hybridization Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
736500-3515 736500-3530
KT736500-3515PK 460.95 CAD
KT736500-3515 KT736500-3530
KIMBLE® KONTES® Hybridization Tubes, DWK Life Sciences
Bottles Hybridisation Bottles/Tubes
For use with hybridization incubators with rotators.
  • Tubes significantly reduce probe volumes
Perform hybridization of multiple Northern, Southern, or Western blots (up to five per tube) in a single tube, using nylon mesh sheets to separate the hybridization membranes.
Ordering information: Complete with 45 mm polypropylene caps and silicone O-rings.