KIMBLE® Gas Washing Bottles, with Fritted Disc, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
657750-1223 657750-2523 657750-2525 657750-5023 657750-5025
KT657750-1223EA 769.38 CAD
KT657750-1223 KT657750-2523 KT657750-2525 KT657750-5023 KT657750-5025
KIMBLE® Gas Washing Bottles, with Fritted Disc, DWK Life Sciences
Bottles Gas Washing Bottles
Tall form gas washing bottle with full length joint with hooks and fritted disc.
  • Available in two porosities: extra coarse (170-220 micron) and coarse (40-60 micron)
  • Supplied complete with hooks and two 2" springs
  • Manufactured from 33 expansion, low extractable borosilicate glass
Bottles contain a fritted disc for more uniform gas distribution throughout the absorbing material.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.