KIMBLE® KIMAX® Gas Washing Bottles, Tall Form, [ST] Stopper, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
15060-125 15060-250 15060-500
89001-568CS 497.16 CAD
89001-568 89001-570 89001-572
KIMBLE® KIMAX® Gas Washing Bottles, Tall Form, [ST] Stopper, DWK Life Sciences
Bottles Gas Washing Bottles
For washing or drying gases.
Gas flows into the bottom of the bottle through the center tube, rises through the washing or drying medium, and exits through the side tube of the stopper. Bottles have [ST] 34/28 stoppers and inlet and outlet ports that accept 0.63 cm (¹/₄") Int.Ø flexible tubing.