Orthoboric acid 99.5-100.5% (by alkalimetry), crystals, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

BX0865-30 BX0865-1 BX0865-3 BX0865-5
CA-BX0865-30EA 6056 CAD
CA-BX0865-30 CABX0865-1 CABX0865-3 CABX0865-5
Orthoboric acid 99.5-100.5% (by alkalimetry), crystals, GR ACS, Supelco®
ortho-Boric acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C (dec., lit.)
Density: 1.49 g/cm3 (23 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric) 99.5-100.5%
Calcium (Ca) 0.002% max
Chloride (Cl) 0.0003% max
Heavy metals (as Pb) 0.0005% max
Insoluble in methanol 0.005% max
Iron (Fe) 0.0001% max
Nonvolatile with methanol 0.05% max
Phosphate (PO4) 0.0005% max
Sulfate (SO4) 0.0005% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR