Benzene ≥99.7%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

BX0212-1 BX0212-6
CABX0212-1CS 2007 CAD
CABX0212-1 CABX0212-6
Benzene ≥99.7%, OmniSolv®, Supelco®
Benzene

For spectrophotometry and chromatography. UV cutoff 280nm. Lot analysis on label.

Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=cae270bfe08ae6ab4b14d9e1775b89a82cfeec1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.7% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 2ppm max
UV Abs. at 280 nm 1.00AU max
UV Abs. at 290 nm 0.15AU max
UV Abs. at 300 nm 0.05AU max
UV Abs. at 330 nm 0.01AU max
UV Abs. at 350 nm 0.005AU max
UV Cut-off 280nm max
Water (H2O) 0.03% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR