Bradford Protein Assay Kit, Solution, Biotechnology Grade, MilliporeSigma
Supplier: MilliporeSigma
2740-1KIT 2740-1KIT
CA2740EA 1184.61 CAD
EM-2740 CA2740
Bradford Protein Assay Kit, Solution, Biotechnology Grade, MilliporeSigma
Assays Protein Assays
Corrosive liquid
Kit contains 1L of Bradford Reagent in a poly bottle, 1.5mL of BSA solution in a 2mL sterile poly tube, and 200mL of 0.15M sodium chloride solution in a poly bottle. All components are packaged in a poly bag.