Bismuth(III) oxide ≥99.9995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

10658-18 10658-30
CAAA10658-18EA 176.96 CAD
CAAA10658-18 CAAA10658-30
Bismuth(III) oxide ≥99.9995% (metals basis), Puratronic®
Bismuth (III) oxide

Powder, 20 mesh. Purity based on metal contaminates only.


Purity: 99.9995% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: -20 Mesh Powder Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Merck Reference: 14,1273 MDL No.: MFCD00003462

Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: ∼1890 °C
Melting Pt: 817 °C
Density: 8.9
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 14,1273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR