Barium carbonate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10645-14 10645-22 10645-36
CAAA10645-14EA 222.78 CAD
CAAA10645-14 CAAA10645-22 CAAA10645-36
Barium carbonate ≥99.997% (metals basis), Puratronic®
Barium carbonate

Powder, 22 mesh. Purity based on metal contaminates only.


Purity: 99.997% CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: -22 Mesh Powder Density: 4.43 Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448

Formula: BaCO₃
Boiling Pt: Dec.
Melting Pt: 811 °C
Density: 4.43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 14,969

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR