Dispensing Bottles, Justrite®
Supplier: Justrite
14009 14011
56610-102EA 112.28 CAD
56610-102 56609-749
Dispensing Bottles, Justrite®
Bottles Dropping Bottles
Polyethylene bottles feature stainless steel ball valve to ensure safer, smoother dispensing of flammable liquids with minimum over-spray. Squeezing bottle produces fine liquid stream. Flexible dispensing tube is self-closing. Stem can be shaped to dispense into areas normally difficult to access.