Azomethine H sodium salt ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A5016-25G A5016-5G
TCA5016-25GEA 447.14 CAD
TCA5016-25G TCA5016-005G
Azomethine H sodium salt ≥97.0% (by titrimetric analysis)
Azomethine H sodium salt

[Spectrophotometric Reagent for B]
CAS Number: 5941-07-1
MDL Number: MFCD00042050
Molecular Formula: C17H13NO8S2
Molecular Weight: 445.39
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulfonic Acid Monosodium Salt

Formula: C₁₇H₁₂NNaO₈S₂
Melting Pt: > 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00042050
CAS Number: 5941-07-1
EINECS: 227-698-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2750acdb3a146e587e23931932c43365067684c6&vid=67ef58aa7755b3e64e1d2a46292c95842d8f999a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR