N-(tert-Butoxycarbonyl)-1,5-diaminopentane ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine
CAS Number: 51644-96-3
MDL Number: MFCD00210020
Molecular Formula: C10H22N2O2
Molecular Weight: 202.30
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 0.97
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Formula:
C₁₀H₂₂N₂O₂ MW: 202.30 g/mol Density: 0.97 (20 °C) |
MDL Number:
MFCD00210020 CAS Number: 51644-96-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...