4-Benzylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A1260-25G A1260-5G
TCA1260-025GEA 685.72 CAD
TCA1260-025G TCA1260-5G
4-Benzylaniline ≥97.0% (by GC, titration analysis)
4-Benzylaniline

CAS Number: 1135-12-2
MDL Number: MFCD00047861
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 44


Other Product Names / Synonyms: 4-Benzylaniline

Formula: C₁₃H₁₃N
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 44 °C
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00047861
CAS Number: 1135-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c083ae586b5959f1e83ff87baa91aadcf59346eb&vid=c083ae586b5959f1e83ff87baa91aadcf59346eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR