O-Acetylricinoleic acid methyl ester ≥80% (by GC)

Supplier: TCI America
Warning

A0879-25ML A0879-500ML
TCA0879-025MLEA 85.32 CAD
TCA0879-025ML TCA0879-500ML
O-Acetylricinoleic acid methyl ester ≥80% (by GC)
O-Acetylricinoleic acid methyl ester

CAS Number: 140-03-4
MDL Number: MFCD00026204
Molecular Formula: C21H38O4
Molecular Weight: 354.53
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Flash Point (°C): 198
Specific Gravity (20/20): 0.94


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: O-Acetylricinoleic Acid Methyl Ester, Methyl (Z)-12-Acetoxy-9-octadecenoate

Formula: C₂₁H₃₈O₄
Boiling Pt: 260 °C
Density: 0.94 (20 °C)
Flash Pt: 198 °C
MDL Number: MFCD00026204
CAS Number: 140-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ca21aba2bbfd5f1070cf9d92e8395877b3982c9&vid=1ca21aba2bbfd5f1070cf9d92e8395877b3982c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR