N-(3-(Trimethoxysilyl)propyl)ethylenediamine ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3-(2-Aminoethylamino)propyltrimethoxysilane, N-(2-Aminoethyl)-3-aminopropyltrimethoxysilane, N-[3-(Trimethoxysilyl)propyl]ethylenediamine
CAS Number: 1760-24-3
MDL Number: MFCD00008173
Molecular Formula: C8H22N2O3Si
Molecular Weight: 222.36
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 143
Specific Gravity (20/20): 1.03
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: N-(2-Aminoethyl)-3-aminopropyltrimethoxysilane, N-(3-Trimethoxysilylpropyl)ethylenediamine
Formula:
H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃ MW: 222.36 g/mol Boiling Pt: 146 °C Melting Pt: 0 Density: 1.03 (20 °C) Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008173 CAS Number: 1760-24-3 EINECS: 217-164-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...