VWR® Timer Plus
Supplier: VWR
VWR®
5045
62379-604EA 115.45 CAD
62379-604
VWR® Timer Plus
Timers Digital Timers
Key chain timer is three timers in one - countdown alarm timer and stopwatch.
  • Traceable®
  • Six-digit LCD display
  • Programmable timer functions
  • VWR® six-year limited warranty
Ideal for catching test results and signaling time intervals and appointments.
Six-digit LCD always shows time in hours, minutes, and seconds. Countdown mode can be programmed in one-minute increments from 100 hours to 1 minute. When display reaches zero, a loud, beeping alarm sounds for 60 seconds. The alarm can be silenced manually. In stopwatch mode, it times up from 1 second to 99 hours and then repeats. The timer has a time-out feature in both countup and stopwatch modes.
Clip it to a button-hole, belt loop, or notebook.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with an eighteen-month continuous-use replaceable silver-oxide battery, keychain, lanyard, and magnet for placing on metal.