Diisopropyl adipate ≥99% (by GC)

Supplier: TCI America

A0654-25ML A0654-500ML
TCA0654-025MLEA 63.2 CAD
TCA0654-025ML TCA0654-500ML
Diisopropyl adipate ≥99% (by GC)
Diisopropyl adipate

CAS Number: 6938-94-9
Molecular Formula: C12H22O4
Molecular Weight: 230.30
Form: Liquid
Color: Colorless
Boiling point (°C): 120
Melting point (°C): -1
Flash Point (°C): 124
Specific Gravity (20/20): 0.97

Formula: C₁₂H₂₂O₄
Boiling Pt: 120 °C
Melting Pt: –1 °C
Density: 0.97
Flash Pt: 124 °C
MDL Number: MFCD00026391
CAS Number: 6938-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f6e037171e73d12d84f2e9c7c3fe6363bf70908&vid=2f6e037171e73d12d84f2e9c7c3fe6363bf70908&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR