8-Aminoquinoline ≥98.0%

Supplier: TCI America
Warning

Synonyms: Quinolin-8-ylamine

A0419-5G A0419-25G A0419-100G
TCA0419-005GEA 94.8 CAD
TCA0419-005G TCA0419-025G TCA0419-100G
8-Aminoquinoline ≥98.0%
8-Aminoquinoline

CAS Number: 578-66-5
MDL Number: MFCD00006809
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 65

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 290
Melting Pt: 64
Density: 1.337 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5
EINECS: 209-427-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef17f43f9e0b836a5d113d336459282397bb35a2&vid=9fa41061723696d4a79ac4c46b01059d28a84c3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR