Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulfonate hydrate ≥80.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: H acid hydrate, 1-Amino-8-naphthol-3,6-disulfonic acid monosodium salt hydrate, 4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid monosodium salt hydrate, 1-Amino-8-naphthol-3,6-disulphonic acid monosodium salt hydrate, 4-Amino-5-hydroxynaphthalene-2,7-disulphonic acid monosodium salt hydrate

A0362-25G A0362-500G
TCA0362-025GEA 82.63 CAD
TCA0362-025G TCA0362-500G
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulfonate hydrate ≥80.0% (by total nitrogen and titration analysis)
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulphonate hydrate

CAS Number: 5460-09-3
MDL Number: MFCD00036444
Molecular Formula: C10H9NO7S2
Molecular Weight: 341.28
Purity/Analysis Method: >80.0% (N,T)
Form: Crystal


Other Product Names / Synonyms: 4-Amino-5-hydroxy-2,7-naphthalenedisulfonic Acid Monosodium Salt, H Acid, 1-Amino-8-naphthol-3,6-disulfonic Acid Monosodium Salt, Monosodium 4-Amino-5-hydroxy-2,7-naphthalenedisulfonate

Formula: C₁₀H₈NNaO₇S₂·xH₂O
MW: 341.28 g/mol (anhy)
MDL Number: MFCD00036444
CAS Number: 343321-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f92feb74c61f35ccea9113572791ff94797d46c&vid=0f92feb74c61f35ccea9113572791ff94797d46c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR