VWR® Dropping Bottles, PTFE
Supplier: VWR International
89026-086 89026-088
89026-086EA 309.9 CAD
89026-086 89026-088
VWR® Dropping Bottles, PTFE
Bottles Dropping Bottles
PTFE dropping bottles are suitable for dispensing and dropping small volumes of reagents and aggressive chemicals.
  • Chemically resistant and inert
  • Tight, leak-free seal
  • Bottle sides are flexible
Bottle sides are flexible to enable user to squeeze out contents. Screw-on cap ensures a tight, leak-free seal.