Ammonia Standard, 1 ml = 1 mg NH₃, 1000 ppm NH₃ (822 ppm N), Ricca Chemical Company
Supplier: Ricca Chemical
R0615000-500A
R0615000-120A
RC61516EA
167.48
CAD
RC61516
RC6154
Ammonia Standard, 1 ml = 1 mg NH₃, 1000 ppm NH₃ (822 ppm N), Ricca Chemical Company
Standards
Cationic Standards
Ammonium Standards
1000ppm as NH3, 822ppm nitrogen.