VWR® Hazardous Location Fridge and Fridge Freezer Combination Units
Supplier: VWR International
77441-862 77441-864
77441-862EA 12547.56 CAD
77441-862 77441-864
VWR® Hazardous Location Fridge and Fridge Freezer Combination Units
Refrigerators
Designed for safety with ATEX and non-sparking components, these units are ideal for keeping you and your substances safe.
  • Microprocessor temperature control
  • ATEX non-sparking components
  • Natural hydrocarbon refrigerants
  • Meet NFPA and OSHA guidelines for hazardous laboratory locations
In the fast-paced world of scientific and industrial laboratories, precision and safety are key. These hazardous location cold storage units are engineered for use in environments where volatile or explosive conditions could exist. Engineered with microprocessor temperature control, they provide exceptional precision, recovery, and maintenance. You’ll also experience the power of natural hydrocarbon refrigerants through energy savings and a greener laboratory footprint—all while enjoying stellar system performance.
Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups A, B, C and D guidelines.
ETL/cETL listed.
Ordering information: Storage baskets and spare shelves are available to purchase separately.
Caution: *Note: Please be aware that these units come without a plug and require hardwiring in conduit, ensuring a secure connection in your high-risk environment. Our freestanding models also require 4 inches of clearance for proper ventilation and safety.