Acetic acid glacial_AcOH 99,7 GC, corrected for water content, AR® ACS Trace_metal, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
M.O.C. Category N
V193-05
V193-45
MKV19305CS
1179.33
CAD
MKV19305
MKV193-45
CAJTV193-44
Acetic acid glacial_AcOH 99,7 GC, corrected for water content, AR® ACS Trace_metal, Macron Fine Chemicals™
Acetic acid
ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 118 Melting Pt: 17 Density: 1.05 g/cm³ (20 °C) Flash Pt: 38.5 Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8 II REACH: 01-2119475328-30 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
IMPORTANT: Material will freeze if stored below 17 °C (63°F). | |
ACS - Assay (by GC, corrected for water) | ≥99.7 % |
ACS - Acetic Anhydride ((CH₃CO)₂O) | ≤0.01 % |
ACS - Chloride (Cl) | ≤0.4 ppm |
ACS - Color (APHA) | ≤10 |
Dilution Test | Passes Test |
ACS - Heavy Metals (by ICP-OES) | ≤5 ppm |
ACS - Residue after Evaporation | ≤8 ppm |
ACS - Substances Reducing Dichromate | Passes Test |
ACS - Substances Reducing Permanganate | Passes Test |
ACS - Sulfate (SO₄) | ≤0.4 ppm |
ACS - Titrable Base (meq/g) | ≤0.0004 |
Trace Impurities - Aluminum (Al) | ≤0.300 ppm |
Trace Impurities - Arsenic (As) | ≤0.050 ppm |
Trace Impurities - Boron (B) | ≤0.200 ppm |
Trace Impurities - Calcium (Ca) | ≤0.300 ppm |
Trace Impurities - Chromium (Cr) | ≤0.200 ppm |
Trace Impurities - Copper (Cu) | ≤0.10 ppm |
Trace Impurities - Gold (Au) | ≤0.300 ppm |
Trace Impurities - Iron (Fe) | ≤0.2 ppm |
Trace Impurities - Lead (Pb) | ≤0.300 ppm |
Trace Impurities - Magnesium (Mg) | ≤0.300 ppm |
Trace Impurities - Manganese (Mn) | ≤0.300 ppm |
Trace Impurities - Nickel (Ni) | ≤0.1 ppm |
Trace Impurities - Potassium (K) | ≤0.300 ppm |
Trace Impurities - Sodium (Na) | ≤0.300 ppm |
Trace Impurities - Tin (Sn) | ≤0.300 ppm |
Trace Impurities - Titanium (Ti) | ≤0.300 ppm |
Trace Impurities - Zinc (Zn) | ≤0.300 ppm |
Appearance (clear, colorless liquid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...