Aniline ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Aminobenzene, Phenylamine
Management of Change (MOC) category = N
3584-04
MACR3584-08
MK358404CS
3823.22
CAD
MK358404
CAMK358404
CAMK358408
Aniline ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
Aniline
ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.
Formula:
C₆H₅NH₂ MW: 93.13 g/mol Boiling Pt: 184 Melting Pt: -6.2 Density: 1.019 g/cm³ (20 °C) Flash Pt: 76 Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS Number: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1 II Merck Index: 13,00661 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (C₆H₅NH₂) (by GC, corrected for water) | ≥99.0 % |
Color (APHA) (at time of packaging) | ≤250 |
Residue after Ignition | ≤0.005 % |
Chlorobenzene (C₆H₅Cl) | ≤0.01 % |
Hydrocarbons | Passes Test |
Nitrobenzene (C₆H₅NO₂) | Passes Test |
NOTE: This reagent darkens to a reddish-brown color on storage. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...