Ammonium sulfate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = NI

MACR3512-06 MACR3512-12 MACR3512-25 MACR3512-28 MACR3512-65 MACR3512-88
CAMK351206CS 1387.76 CAD
CAMK351206 CAMK351212 CAMK351225 CAMK351228 CAMK351265 CAMK351288
Ammonium sulfate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Ammonium sulphate

Granular.

Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.77 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥99.0 %
ACS - Residue on Ignition ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤3 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - Phosphate (PO₄) ≤3 ppm
ACS - Iron (Fe) ≤5 ppm
Heavy Metals (as Pb) ≤3 ppm
Arsenic (As) ≤0.200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR