VWR® Compact Anemometer/Thermometer
Supplier: VWR
VWR®
VC4331 4331
33500-096EA 760.23 CAD
10754-710 33500-096
VWR® Compact Anemometer/Thermometer
Anemometers
Instrument measures air velocity and temperature with simple, one-hand operation.
  • Traceable®
  • Compact Design
Readings are updated instantaneously. The memory function recalls highest and lowest readings; the hold key freezes the display for capturing a reading in a difficult location; the average key automatically calculates mean value for up to eight readings. Rugged chemical- and shock-resistant ABS plastic case ensures years of reliable service even in severe lab, plant, or outdoor environments. Extra-large 1.3 cm (1/2") LCD digits are legible from 3m (10').
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with RS-232 serial output jack, 9V alkaline battery, instructions, and carrying case.