Ammonia solution 27,0 - 30,0% NF, Multi-Compendial for cleaning and processing, for biochemistry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

Management of Change (MOC) category = R

BAKR9736-05 BAKR9736-09
CAJT9736-5CS 3478.92 CAD
CAJT9736-5 CAJT9736-9
Ammonia solution 27,0 - 30,0% NF, Multi-Compendial for cleaning and processing, for biochemistry, J.T.Baker®
Ammonia solution

pH adjusters are used to adjust the pH of formulations. This category includes acids that are notoriously difficult to source as truly cGMP-produced and amino acids that most companies do not fully test for EP, BP and JP. Avantor provides all that and more with our cGMP-produced acids and mutli-compendial amino acids.

Formula: NH₃
Boiling Pt: –33 °C (1013 hPa)
Melting Pt: –77 °C
Density: 0.77 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 231-635-3
UN: 2672
ADR: 8,
Merck Index: 13,00492

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Nonvolatile Residue ≤0.05 %
NF - Readily Oxidizable Substances Passes Test
NF - Assay (NH₃)(NF) 27.0 - 31.0 %
EP/BP - Identification A (EP/BP) Passes Test
EP/BP - Identification B (EP/BP) Passes Test
EP/BP - Identification C (EP/BP) Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Oxidizable Substances Passes Test
EP/BP - Pyridine and Related Substances ≤2 ppm
EP/BP - Residue on Evaporation (mg/L) ≤20
EP/BP - Carbonates (CO₃) ≤60 ppm
EP/BP - Chlorides (Cl) ≤1 ppm
EP/BP - Sulfates (SO₄) ≤5 ppm
EP/BP - Iron (Fe) ≤0.25 ppm
EP/BP - Assay (NH₃)(EP/BP) 25.0 - 30.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR