Acetic acid glacial ≥99.9%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

Management of Change (MOC) category = N

BAKR9524-00 BAKR9524-33
CAJT9524-0CS 1288.6 CAD
CAJT9524-0 CAJT9524-33
Acetic acid glacial ≥99.9%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Acetic acid

For trace metal analysis. Certificate of lot analysis provided.

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
ACS - Assay (by GC, corrected for water) ≥99.9 %
ACS - Acetaldehyde ≤0.005 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
ACS - Color (APHA) ≤10
Dilution Test Passes Test
ACS - Residue after Evaporation ≤5 ppm
ACS - Solubility in H₂O Passes Test
ACS - Specific Gravity at 20°/20°C ≥1.048
ACS - Substances Reducing Dichromate Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Titrable Base (meq/g) ≤0.0004
Chloride (Cl) ≤0.5 ppm
Phosphate (PO₄) ≤1 ppm
Sulfate (SO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤20.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤50 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤5.0 ppb
Trace Impurities - Calcium (Ca) ≤100.0 ppb
Trace Impurities - Chromium (Cr) ≤50 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤50.0 ppb
Heavy Metals (as Pb) ≤300.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤50.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Manganese (Mn) ≤5.0 ppb
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤200.0 ppb
Trace Impurities - Silicon (Si) ≤50 ppb
Trace Impurities - Silver (Ag) ≤5.0 ppb
Trace Impurities - Sodium (Na) ≤500.0 ppb
Trace Impurities - Strontium (Sr) ≤5.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Titanium (Ti) ≤300.0 ppb
Trace Impurities - Zinc (Zn) ≤10.0 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR