Protektive Pak® ESD Shipping Boxes, Desco Industries
Supplier: Protektive Pak
37061 37000 37008 37001 37004 37003 37006
30000-062EA 32 CAD
30000-062 30000-798 30000-800 30000-802 30000-804 30000-806 30000-812
Protektive Pak® ESD Shipping Boxes, Desco Industries
Boxes ESD-Safe Boxes
Shipping boxes are made of impregnated corrugated material that provides greater durability than coated or printed material
Small component shippers are an effective means of shipping or storing small ESD-sensitive items. Interior features pink static-dissipative polyurethane foam laminated to the top and removable black conductive crosslink polyethylene foam in the bottom.
Circuit board shippers are an effective means of shipping or storing small ESD-sensitive circuit boards and other small items. Interior features convoluted static-dissipative (<1011ohms/sq.) pink polyurethaneMore Product Information