Acetone ≥99.4% (by GC, corrected for water content), ULTRA RESI-ANALYZED™, ACS Reagent for gas chromatography, for pesticide residue analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Management of Change (MOC) category = NI

BAKR9254-02 BAKR9254-03
CAJT9254-2CS 2880.64 CAD
CAJT9254-2 CAJT9254-3
Acetone ≥99.4% (by GC, corrected for water content), ULTRA RESI-ANALYZED™, ACS Reagent for gas chromatography, for pesticide residue analysis, J.T.Baker®
Acetone

BAKER ULTRA RESI-ANALYZED® Reagent—ultra high-purity solvents and salts developed for organic residue extraction/concentration procedures.


These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors. Advanced stabilizer systems are incorporated to ensure solvent stability and superior performance. Solvents are suitable for exacting EPA sample cleanup protocols.

Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.4 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Reducing Permanganate Passes Test
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.6
Water (H₂O) ≤0.5 %
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR