Acetonitrile ≥99.9% (by GC), BAKER ANALYZED® HPLC, Ultra Gradient for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Management of Change (MOC) category = N

BAKR9017-02 BAKR9017-03 9017-33
CAJT9017-2CS 2310.09 CAD
CAJT9017-2 CAJT9017-3 CAJT9017-33 JT9017-33 CAJT9017-69 CAJT9017-88 CAJT9017-S1
Acetonitrile ≥99.9% (by GC), BAKER ANALYZED® HPLC, Ultra Gradient for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Acetonitrile

Packaged under nitrogen. For high performance liquid chromatography. UV cutoff 190nm.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤190
Gradient Elution Test (a.u.) - 254 nm ≤0.0005
Gradient Elution Test (a.u.) - 210 nm ≤0.002
Density (g/mL) at 25°C 0.775 - 0.780
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (CH₃CN) (by GC) ≥99.9 %
Appearance Passes Test
Color (APHA) ≤10
Fluorescence Trace Impurities, measured as Quinine Base - Fluorescence detection (PAH) ≤0.5 ppb
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.8
Titrable Base (µeq/g) ≤0.6
Water (by KF, coulometric) ≤100 ppm
Carbonyl Compounds (as Acetone) ≤25.0 ppb
Filtered through a 0.2 micron filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR