Acetone ≥99.5% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Management of Change (MOC) category = N

BAKR9010-03 9010-01
CAJT9010-3CS 2001.07 CAD
CAJT9010-3 JT9010-1
Acetone ≥99.5% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Acetone

For spectrophotometry. Contains 0.5% water. Lot analysis on label.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.5 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0005 %
ACS - Solubility in H₂O Passes Test
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Titrable Base (meq/g) ≤0.0006
ACS - Aldehydes (as HCHO) ≤0.002 %
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
ACS - Methanol (CH₃OH)(by GC) ≤0.05 %
ACS - Substances Reducing Permanganate Passes Test
ACS - Water (H₂O) ≤0.5 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm ≤0.010
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm ≤1.00
Product Information (not specifications):
Density (g/mL) at 25 °C (typical)........ 0.7857
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm:
2.5 - 3.2; 3.6 - 5.5; 8.6 - 8.9; 9.4 - 10.7; 11.4 - 12


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR