Acetone ≥99.5% NF, FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
Management of Change (MOC) category = R
BAKR9008-R
9008-03
CAJT9008-REA
5618.46
CAD
CAJT9008-R
JT9008-3
Acetone ≥99.5% NF, FCC, J.T.Baker®
Acetone
Formula:
CH₃COCH₃ Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets F.C.C. Requirements | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Benzene | ≤2 ppm |
NF - Specific Gravity at 25°/25°C | ≤0.788 |
NF - Nonvolatile Residue | ≤10 ppm |
NF - Readily Oxidizable Substances | Passes Test |
NF - Water (H₂O) | ≤0.5 % |
NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis) | 99.5 - 100.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Solubility in H₂O | Passes Test |
FCC - Substances Reducing Permanganate | Passes Test |
FCC - Acidity (as CH₃COOH) | ≤0.002 % |
FCC - Aldehydes (as HCHO) | ≤0.002 % |
FCC - Alkalinity (as NH₃) | ≤10 ppm |
FCC - Lead (Pb) | ≤1 mg/kg |
FCC - Methanol (CH₃OH) | ≤0.05 % |
FCC - Phenols | Passes Test |
FCC - Refractive Index at 20°C | 1.358 - 1.360 |
FCC - Nonvolatile Residue | ≤10 ppm |
FCC - Water (H₂O) | ≤0.5 % |
FCC - Distilling Range | ≤1 °C |
FCC - Initial Boiling Point | |
FCC - Dry Point | |
Class 1, 2 and 3 residual solvents are likely to be present. Benzene (Class 1) is reported on each lot and is below the USP/NF limit concentration. Methanol and Toluene (Class 2) are both below their | |
Option 1 limits. All Class 3 solvents are less than 0.5%. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...