Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
BAKR9006-R
BAKR9006-01
BAKR9006-22
BAKR9006-03
BAKR9006-33
BAKR9006-05
BAKR9006-06
BAKR9006-07
CAJT9006-REA
3630.48
CAD
CAJT9006-R
CAJT9006-1
CAJT9006-22
CAJT9006-3
CAJT9006-33
CAJT9006-5
CAJT9006-6
CAJT9006-7
CAJT9006-ZT
Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Acetone
The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.
- Produced using proven distillation methods and rigid testing protocols
- Statistical Process Control used in purification and packaging to ensure lot-to-lot consistency
- Statistical Quality Control and suitability-for-use tests to ensure product performance
Formula:
CH₃COCH₃ Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay ((CH₃)₂CO) (by GC, corrected for water) | ≥99.5 % |
Color (APHA) | ≤10 |
Residue after Evaporation | ≤0.001 % |
Solubility in H₂O | Passes Test |
Titrable Acid (meq/g) | ≤0.0003 |
Titrable Base (meq/g) | ≤0.0006 |
Aldehydes (as HCHO) | ≤0.002 % |
Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) | ≤0.05 % |
Methanol (CH₃OH)(by GC) | ≤0.05 % |
Heavy Metals (as Pb) | ≤1 ppm |
Substances Reducing Permanganate | Passes Test |
Water (H₂O) | ≤0.5 % |
Copper (Cu) | ≤0.1 ppm |
Iron (Fe) | ≤0.1 ppm |
Nickel (Ni) | ≤0.1 ppm |
Date Received: __ __ /__ __ /__ __ | |
Date Opened: __ __ /__ __ /__ __ | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...