Acetone ≥99.3% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR9002-02 BAKR9002-03 BAKR9002-XV
CAJT9002-2CS 1154.51 CAD
CAJT9002-2 CAJT9002-3 CAJT9002-33 CAJT9002-88 CAJT9002-YE CAJT9002-XV
Acetone ≥99.3% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Acetone

Packaged under nitrogen. Filtered through a 0.2µm filter to remove particulate matter. UV cutoff 330nm. Lot analysis on label.


  • Packaged under nitrogen
  • Filtered through a 0.2 µm filter to remove particulate matter

Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤330
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay ((CH₃)₂CO)(by GC) ≥99.3 %
Residue after Evaporation ≤2.0 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.6
Water (H₂O) ≤0.4 %
Filtered through a 0.2 micron filter.
Packaged under Nitrogen.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR