Ammonia solution 29%, Finyte®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Ammonia aqueous
, Ammonium hydroxide solution
, Ammonia water
, Ammonium hydroxide
5817-09
BAKR5817-09
5817-CN
CAJT5817-9EA
1704.71
CAD
JT5817-9
CAJT5817-9
CAJT5817-CN
Ammonia solution 29%, Finyte®, J.T.Baker®
Ammonia solution
Formula:
NH₃ MW: 35.05 g/mol Boiling Pt: 32…37.7 °C (1013 hPa) Melting Pt: –60…–57.5 °C Density: 0.88…0.91 g/cm³ (20 °C) |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 UN: 2672 ADR: 8,III |
Specification Test Results
For Microelectronic Use | |
Appearance | Passes Test |
Assay (as NH₃) | 28.0 - 30.0 % |
Color (APHA) | ≤10 |
Residue after Ignition | ≤3 ppm |
Carbon Dioxide (CO₂) | ≤10 ppm |
Substances Reducing Permanganate | Passes Test |
Chloride (Cl) | ≤0.2 ppm |
Nitrate (NO₃) | ≤0.4 ppm |
Phosphate (PO₄) | ≤0.10 ppm |
Total Sulfur (as SO₄) | ≤0.2 ppm |
Trace Impurities - Aluminum (Al) | ≤10.0 ppb |
Arsenic and Antimony (as As) | ≤5.0 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤10.0 ppb |
Trace Impurities - Boron (B) | ≤10.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤5.0 ppb |
Trace Impurities - Calcium (Ca) | ≤20 ppb |
Trace Impurities - Chromium (Cr) | ≤5.0 ppb |
Trace Impurities - Cobalt (Co) | ≤1.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤10.0 ppb |
Trace Impurities - Gold (Au) | ≤10.0 ppb |
Trace Impurities - Iron (Fe) | ≤10.0 ppb |
Trace Impurities - Lead (Pb) | ≤10.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤10.0 ppb |
Trace Impurities - Manganese (Mn) | ≤5.0 ppb |
Trace Impurities - Molybdenum (Mo) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤5.0 ppb |
Trace Impurities - Niobium (Nb) | ≤10.0 ppb |
Trace Impurities - Potassium (K) | ≤10.0 ppb |
Trace Impurities - Silicon (Si) | ≤10.0 ppb |
Trace Impurities - Silver (Ag) | ≤5.0 ppb |
Trace Impurities - Sodium (Na) | ≤10.0 ppb |
Trace Impurities - Strontium (Sr) | ≤10.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤10.0 ppb |
Trace Impurities - Tin (Sn) | ≤10.0 ppb |
Trace Impurities - Titanium (Ti) | ≤10.0 ppb |
Trace Impurities - Vanadium (V) | ≤10.0 ppb |
Trace Impurities - Zinc (Zn) | ≤10.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤10.0 ppb |
Particle Count at point of fill - 0.5 µm and greater | ≤150 par/ml |
Particle Count at point of fill - 0.2 µm and greater, For Information Only | |
Particle Count at point of fill - 1.0 µm and greater | ≤25 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...