Ammonia solution 5 N, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

M.O.C. Category N

BAKR5604-07 BAKR5604-02
CAJT5604-7EA 1385.49 CAD
CAJT5604-7 CAJT5604-2
Ammonia solution 5 N, BAKER ANALYZED® Volumetric, J.T.Baker®
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Density: 0.90 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 4.95 - 5.05
Standardization at 25°C traceable to NIST Standard Reference Material.
Normality should be confirmed at the time of use because Ammonium
Hydroxide may vaporize thus reducing its normality.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR