KODAK® BioMax® Light Autoradiography Film, Carestream Health
Supplier: Kodak
1917012 8689358
CAIB-1917012CS 26595 CAD
CAIB-1917012 CAIB-8689358 CAIB-8761520 IB-1788207 IB-8194540 IB-8689358
KODAK® BioMax® Light Autoradiography Film, Carestream Health
Autoradiography Films Films
Double emulsion film provides maximum clarity and sensitivity for detection of chemiluminescent labeled samples
Also suitable for direct and intensifying screen exposures with commonly used isotopes. However, the film provides lower sensitivity and requires longer exposure times compared to BioMax® MR, BioMax® MS, and X-OMAT® AR films for radioisotope detection. Suitable for manual and automated processing.
Produces the highest signal-to-noise ratio of any KODAK® film for detection of chemiluminescence.
Ordering information: Available non-interleaved or individually wrapped.