Jackson Safety®G 40 Latex Coated Gloves, Kimberly-Clark
Supplier: Microflex
97270 97271 97272 97273 97274
CA89011-794CS 660.36 CAD
CA89011-794 CA89011-796 CA89011-798 CA89011-800 CA89011-802
Jackson Safety®G 40 Latex Coated Gloves, Kimberly-Clark
Gloves
General-purpose, latex-coated, poly-cotton gloves provide a secure grip as well as great comfort and dexterity.
  • Now washable
  • Processed to yield lower latex proteins
  • Gloves feature a puncture rating of EN 388 (ACTP) 2142
  • Eco-friendly packaging, with no separate insert, no printed bags, and less waste
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.
Durable, silicone-free gloves are ideal for industrial use, such as masonry applications and shipping and receiving. They offer improved latex coating lamination strength, wider palm widths for more generous sizing, and a 100% polyester liner for better adhesion to coating. Gloves also provide an improved heat transfer stamp, with ratings visible even after glove use (EN407 Contact Heat Level 2).