Nalgene® Baffled Culture Flasks, Polycarbonate, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
4110-0500 4110-0250 4110-2000 4110-1000
29151-702CS 666.13 CAD
29151-702 29151-700 29151-706 29151-704
Nalgene® Baffled Culture Flasks, Polycarbonate, Thermo Scientific
Flasks Cell Culture Flasks
For E coli and S. cerevisiae at room temperature, polycarbonate flasks produce growth curves equivalent to those of glass flasks.
Baffle design supplies the mixing, aeration, and gas exchange needed for effective cell culture, fermentation, and genetic engineering procedures. Molded neck ridge acts as a finger stop for consistent sterile technique. The 500 and 1000mL sizes have a straight neck design. Autoclavable.