Ammonium hydrogen carbonate 21,30 - 21,73%, powder, BAKER ANANYZED™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
BAKR3003-01
BAKR3003-05
CAJT3003-1EA
133.78
CAD
CAJT3003-1
CAJT3003-5
Ammonium hydrogen carbonate 21,30 - 21,73%, powder, BAKER ANANYZED™
Ammonium hydrogen carbonate
Powder.
Formula:
NH₄HCO₃ Melting Pt: 35 °C Density: 1.57 g/cm³ (20 °C) |
MDL Number:
MFCD00012138 CAS Number: 1066-33-7 EINECS: 213-911-5 Merck Index: 13,00497 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (as NH₃) | 21.30 - 21.73 % |
pH of 5% Solution at 25°C | 7.0 - 8.0 |
Nonvolatile Matter | ≤0.02 % |
Insoluble Matter | ≤0.005 % |
Nitrate (NO₃) | ≤0.002 % |
Sulfur Compounds (as SO₄) | ≤0.002 % |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...