L(+)-Ascorbic acid 99.0-100.5%, fine-granular USP, FCC 20 - 80 Mesh, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

Management of Change (MOC) category = R

BAKR0938-05 BAKR0938-07 BAKR0938-08 0938-08
CAJT0938-5EA 743.44 CAD
CAJT0938-5 CAJT0938-7 CAJT0938-8 JT0938-8
L(+)-Ascorbic acid 99.0-100.5%, fine-granular USP, FCC 20 - 80 Mesh, J.T.Baker®
L(+)-Ascorbic acid

Fine granules. 20 to 80 mesh.


  • Food GMP manufactured product

Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553 °C (1013 hPa)
Melting Pt: 190…192 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (C₆H₈O₆) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 20.5 - 21.5 Degree
USP - Residue on Ignition ≤0.1 %
FCC - Assay (C₆H₈O₆) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Specific Rotation [Å]²⁵ ^D (+) 20.5 - 21.5 Degree
FCC - Residue on Ignition ≤0.1 %
Arsenic (As) ≤3 ppm
Mesh - Thru U.S. No. 20 Sieve ≥100 %
Mesh - Thru U.S. No. 30 Sieve ≥95 %
Mesh - Thru U.S. No. 80 Sieve ≤20 %
Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3
Solvents are likely to be present. Residual Class 2 Solvents are below
Option 1 and the residual concentration of all Class 3 Solvents is
<0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR