Glass Dropping Bottles, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W211734 W211735 W211754 W211755
14216-230CS 219.97 CAD
14216-230 14216-232 14216-234 14216-236
Glass Dropping Bottles, Wheaton®, DWK Life Sciences
Bottles Dropping Bottles
With glass hood stopper.
Channel in stopper allows drop-by-drop delivery of contents when bottle is tipped. A partial turn of the stopper seals bottle. Available in clear and amber glass. USD Type III Glass.