Protektive Pak® ESD Nesting Totes, Desco Industries
Supplier: Protektive Pak
37620 37621 37622 37623
30000-820EA 30.98 CAD
30000-820 30000-822 30000-824 30000-826
Protektive Pak® ESD Nesting Totes, Desco Industries
Boxes ESD-Safe Boxes
Used to transport or store ESD-sensitive items such as circuit boards
Totes are made of impregnated, corrugated cardboard. A buried shielding layer minimizes sloughing and rub-off contamination. Feature a static-dissipative surface of 107–109ohms/sq. The conductive handles provide a reliable path-to-ground. Made from 100% recycled material and are 100% recyclable.
Order Now
SPECIFICATIONS
Environmentally Preferable