Dropping Bottles, Amber, Kimble®, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
15040G-30 15040P-60 15040P-30 15040G-60
16199-031CS 81.75 CAD
16199-031 16199-039 16199-034 16199-037
Dropping Bottles, Amber, Kimble®, DWK Life Sciences
Bottles Dropping Bottles
ASTM E438, Type III amber glass protects light-sensitive chemicals.
Supplied with 20-400 GPI screw cap, bulb and dropper assembled on the bottle. Dropper is available in polyethylene or USP Type III glass.